Order Entry
Mexico
Orders LinkContactUsLinkComponent
Agar for bacteriology

Agar for bacteriology

Supplier:   VWR
Agar for bacteriology
Supplier:  VWR
Certificates
Agar for bacteriology
Agar for bacteriology

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A solidifying agent used in bacteriology. Agar consists of unbranched polysaccharides of galactose subunits derived from algae or seaweed.

  • Agar for the growth of bacteria and fungi
  • Typically used at concentrations of 1-2% in solid media preparation
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1

Documents

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 kg
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $4,686.14
    $4,686.14
    Log in for availability
    Case de 4 (1UN)
    Case de 4 (1UN) - $5,107.90
    $5,107.90
    Each (1UN)
       $4,686.14

    Case de 4 (1UN)
       $5,107.90

    Supplier Number: J637-1KG
  • 2.5 kg
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $9,706.08
    $9,706.08
    Each (1UN)
       $9,706.08

    Supplier Number: J637-2.5KG
  • 10 kg
    Log in for availability
    Log in for availability
    Each (10kg)
    Each (10kg) - $32,457.48
    $32,457.48
    Each (10kg)
       $32,457.48

    Supplier Number: J637-10KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.