Order Entry
Mexico
Orders LinkContactUsLinkComponent
Sodium hydroxide, pellets, Proteomics Grade

Sodium hydroxide, pellets, Proteomics Grade

Corrosive
Supplier:   VWR
Sodium hydroxide, pellets, Proteomics Grade
Supplier:  VWR
Certificates
Sodium hydroxide, pellets, Proteomics Grade

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390°C
  • Boiling Pt:1390 °C
  • Melting Pt:318°C
  • Melting Pt:318 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Chloride:
    0.005%
  • DNase:
    none detected
  • Heavy Metals:
    0.002 %
  • Iron:
    = 0.001 %
  • Mercury:
    = 0.1 ppm
  • Nickel:
    = 0.001 %
  • Nitrogen Compounds:
    = 0.001 %
  • Phosphate:
    = 0.001 %
  • Potassium:
    = 0.02 %
  • Protease:
    none
  • Purity:
    = 97 %
  • RNase:
    none detected
  • Sodium Carbonate:
    = 1 %
  • Sulfate:
    = 0.003%
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    500 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $2,353.09
    $2,353.09
    Log in for availability
    Case de 12 (500g)
    Case de 12 (500g) - $2,564.86
    $2,564.86
    Each (500g)
       $2,353.09

    Case de 12 (500g)
       $2,564.86

    Supplier Number: M137-500G
  • HDPE bottle
    1 kg
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $1,919.89
    $1,919.89
    Log in for availability
    Case de 4 (1kg)
    Case de 4 (1kg) - $2,092.68
    $2,092.68
    Each (1kg)
       $1,919.89

    Case de 4 (1kg)
       $2,092.68

    Supplier Number: M137-1KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.