Order Entry
Mexico
Orders LinkContactUsLinkComponent
Urea, Proteomics Grade

Urea, Proteomics Grade

HDPE bottle, 1 kg
Catalog # 97063-798
Supplier:   VWR
Supplier #:  M123-1KG
Urea, Proteomics Grade
Catalog # 97063-798
Supplier:  VWR
CAS Number:  57-13-6
Urea, Proteomics Grade
Urea, Proteomics Grade

HDPE bottle, 1 kg

CAS Number:  57-13-6
Price
$3,883.08
Log in for availability
Each (1kg) Case de 4 (1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the acti

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33

Documents

Specifications

Specification Test Results

  • Abs.@280nm (5M, Water):
    0.05
  • Biuret Test:
    Pass
  • Chloride:
    = 0.0005 %
  • Cyanate:
    = 0.002 %
  • DNase:
    none detected
  • Heavy Metals (as Pb):
    = 0.0005 %
  • Insolubles:
    0.005 %
  • Iron:
    = 0.0001 %
  • Melting Point:
    132°C - 135°C
  • Protease:
    none
  • Purity:
    = 99.5 %
  • RNase:
    none detected
  • Solubility (5M, Water):
    Pass

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.