


Acrylamide (monomer) for biochemistry
97064-866
: VWR
:
- Size:500 g
- Cat. no.:97064-866
- Supplier No.:M120-500G
- Conductivity (40%, Water):5 umhos
- DNase:none detected
- Electrophoresis:Pass
- Free Acrylic Acid:0.001 %
- Appearance:White free-flowing, uniform crystals
- Insolubles:0.005 %
- Iron:0.0001 %
- Lead:0.0001 %
- Magnesium:0.0001 %
- Melting Point:84°C - 86°C
- Moisture (KF):0.2 %
- pH (10%, 0.1M NaCl) @25°C:6.5 %
- Protease:none detected
- Purity:= 99.9%
:
:71.08 g/mol
:125 °C
:125°C
:84.5 °C
:84.5°C
:1.122 g/cm³ (20 °C)
:138 °C
:138 °C
:Refrigerator
:201-173-7
:3426
:6.1,III
:GHS06
:GHS08
:GHS07
:79-06-1
:C₃H₅NO
:MFCD00008032