D-(-)-Sorbitol ≥96%, high purity
Supplier: VWR
About this item
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:98°C - 100°C
- Melting Pt:98…100 °C
- Density:1.524 g/cm³ (20 °C)
- Flash Pt:≥ 100 °C
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
2 Options Available Below
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:98°C - 100°C
- Melting Pt:98…100 °C
- Density:1.524 g/cm³ (20 °C)
- Flash Pt:≥ 100 °C
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Documents
Certificates
Search by Lot NumberPall Laboratory gDNA Purification from Cultured Yeast Cells, NAB
Product Literature
Specifications for Product Family
Specification Test Results
- Arsenic:0.0003 %
- Chloride:0.005 %
- Heavy Metals:0.001 %
- Moisture (KF):= 1 %
- Purity (Anhydrous):= 96 %
- Residue after Ignition:0.1 %
- Sulfate:0.01 %
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- HDPE bottle500 gEach (500g)$1,361.89Case de 12 (500g)$1,484.46Supplier Number: 0691-500G
- Supplier Number: 0691-5KG
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



