Order Entry
Mexico
ContactUsLinkComponent
Nitric acid 68,0 - 70,0% ACS
Nitric acid 68,0 - 70,0% ACS
Catalog # 97064-628
Supplier:  VWR
CAS Number:  
undefined
Nitric acid 68,0 - 70,0% ACS
Catalog # 97064-628
Supplier:  VWR
Supplier Number:  0587-100ML
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Bottle, amber
  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. no.:
    97064-628
  • Supplier No.:
    0587-100ML

Specifications

Specification Test Results

  • Arsenic:
    0.01 ppm
  • Chloride:
    = 0.5 ppm
  • Color (APHA):
    = 10
  • Heavy Metals:
    = 0.2 ppm
  • Iron:
    = 0.2 ppm
  • Purity:
    68% -70%
  • Residue on Ignition:
    = 5 ppm
  • Sulfate:
    = 1 ppm

Specification Test Results

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
Melting Pt:-42
EINECS:231-714-2
UN:2031
 :GHS03
 :GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:87
Density:1.4 g/cm³ (20 °C)
ADR:8 II
Merck Index:13,06610