(±)-1,2-Propanediol, high purity
Supplier: VWR
About this item
Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
- CAS Number:57-55-6
- Formula:CH₃CH(OH)CH₂OH
- MDL Number:MFCD00064272
- Molecular Weight:76.1 g/mol
- Merck Index:13,07947
- Boiling Pt:187.6 °C (1013 hPa)
- Melting Pt:-59 °C
- Melting Pt:–59 °C
- Density:1.04 g/cm³ (20 °C)
- Flash Pt:99 °C
- Storage Temperature:Ambient
- EINECS:200-338-0
2 Options Available Below
Product Details & Documents
Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
- CAS Number:57-55-6
- Formula:CH₃CH(OH)CH₂OH
- MDL Number:MFCD00064272
- Molecular Weight:76.1 g/mol
- Merck Index:13,07947
- Boiling Pt:187.6 °C (1013 hPa)
- Melting Pt:-59 °C
- Melting Pt:–59 °C
- Density:1.04 g/cm³ (20 °C)
- Flash Pt:99 °C
- Storage Temperature:Ambient
- EINECS:200-338-0
Certificates
Search by Lot NumberSpecifications for Product Family
Specification Test Results
- Arsenic:= 0.0003 %
- Chloride:= 0.007 %
- Heavy Metals:= 0.0005 %
- Purity:= 99.5 %
- Residue on Ignition:= 0.01 %
- Sulfate:= 0.006 %
- Water (KF):= 0.2 %
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



