Order Entry
Mexico
ContactUsLinkComponent
Sodium thiosulphate
Sodium thiosulphate
Catalog # 97061-554
Proveedor:  VWR
Número CAS:  
undefined
Sodium thiosulphate
Catalog # 97061-554
Proveedor:  VWR
Supplier Number:  0393-500G
Número CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Especificaciones

  • Pack type:
    HDPE bottle
  • Size:
    500 g
  • Cat. no.:
    97061-554
  • Supplier No.:
    0393-500G

Especificaciones

Resultados de prueba de especificación

  • Heavy Metals (as Pb):
    10 ppm
  • Insolubles:
    0.4 %
  • Iron:
    0.002 %
  • Purity:
    98.3 %
  • Sulfide:
    0.0002 %

Resultados de prueba de especificación

Acerca de este artículo

 :
 :158.11 g/mol
Índice Merck:12,08844
Punto de ebullición:100 °C (1013 hPa)
Punto de ebullición:100 °C (1013 hPa)
Punto de fusión:48 °C
Densidad:1.66 g/cm³ (20 °C)
Temperatura de almacenamiento:Ambient
EINECS:231-867-5
Número CAS:7772-98-7
Fórmula:Na₂S₂O₃
Número MDL:MFCD00003499