Silver nitrate ACS
Bottle, amber, 25 g
Supplier: VWR
Supplier #: 0377-25G
Jump to:
Product Details & Documents
Staining protein after electrophoretic separation. Also used to determine chloride ions in solution.
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (1013 hPa)
- Melting Pt:210 °C
- Density:4.35 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Documents
Certificates
Search by Lot NumberVWR GHS Compliance Letter (US)
Additional Product Details
VWR Life Science AMRESCO Protein Analysis and Detection
Product Literature
Specifications
Specification Test Results
- Appearance (ACS):Colorless or white solid.
- Chloride:= 0.0005%
- Clarity of Solution:Pass
- Copper:= 0.0002%
- Free Acid Test:Pass
- Iron:= 0.0002%
- Lead:= 0.001%
- Purity:= 99%
- Substances not Precipitated by HCl:= 0.01%
- Sulfate:= 0.002%
Specifications
- Pack type:Bottle, amber
- Size:25 g



