Order Entry
Mexico
ContactUsLinkComponent
 
Magnesium sulphate
Catalog # 97061-438
Supplier:  VWR
CAS Number:  
undefined
Magnesium sulphate
Catalog # 97061-438
Supplier:  VWR
Supplier Number:  0338-500G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    500 g
  • Cat. no.:
    97061-438
  • Supplier No.:
    0338-500G

Specifications

Specification Test Results

  • Moisture:
    0.2 %
  • Purity:
    98 %

Specification Test Results

About this item

 :
 :120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110