Methanol ≥99.8%
Catalog # 97065-052
Supplier: VWR
CAS Number:
Specifications
- Pack type:HDPE bottle
- Size:4 L
- Cat. no.:97065-052
- Supplier No.:0323-4L
Specifications
Specification Test Results
- Moisture (KF):= 0.1 %
- Purity:= 99.8 %
- Specific Gravity:0.783 - 0.791
Specification Test Results
About this item
:
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C
Boiling Pt:64.6°C
Melting Pt:–98 °C
Melting Pt:-98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 Subrisk 6.1,II
REACH:01-2119433307-44
:GHS06
:GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595