


Sodium dodecyl sulphate (SDS) for biochemistry
Supplier: VWR
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Merck Index:14,08636
Melting Pt:204°C-207°C
Density:1.1 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,III
:GHS06
:GHS02
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Abs.@260nm (3%, Water):0.1
- Abs.@280nm (3%, Water):= 0.1
- Chloride:= 0.1%
- Copper:= 0.0005%
- DNase:none detected
- Heavy Metals (as Pb):= 0.0005%
- Iron:= 0.0001%
- Phosphate:= 0.0005%
- Protease:none detected
- Purity:= 99%
- RNase:none detected
- Solubility (10%, Water):Pass
Specification Test Results
Related Information
Certificates
Search by Lot NumberVWR GHS Compliance Letter (US)
Additional Product Details
VWR Life Science Biochemical and Molecular Biology Reagents
Product Literature
Frequently Bought Together