Order Entry
Mexico
ContactUsLinkComponent
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%, Ultra Pure Grade for biochemistry
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%, Ultra Pure Grade for biochemistry
Catalog # 97061-226
Supplier:  VWR
CAS Number:  
undefined
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%, Ultra Pure Grade for biochemistry
Catalog # 97061-226
Supplier:  VWR
Supplier Number:  0220-25G
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    25 g
  • Environmentally Preferable:
  • Cat. no.:
    97061-226
  • Supplier No.:
    0220-25G

Specifications

Specification Test Results

  • A250/A260:
    0.75 - 0.81
  • A280/A260:
    0.14 - 0.18
  • AMP & ADP:
    = 0.5 %
  • Em (259nm,Phosphate Buffer,pH 7.0):
    = 14700
  • Heavy Metals:
    = 0.003 %
  • Moisture (KF):
    10 %
  • Purity:
    98 %
  • Sodium (Flame Photom):
    7 % - 8 %
  • Solubility (5%, Water):
    pass

Specification Test Results

About this item

Synonyms: (((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, disodium salt trihydrate, 5'-ATP·2Na·3H2O, Adenosine-5'-triphosphoric acid disodium salt trihydrate, ATP, 2Na 3H2O, ATP disodium trihydrate
 :
 :605.19 g/mol
Storage Temperature:Freezer
EINECS:213-579-1
CAS Number:51963-61-2
 :987-65-5
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MDL Number:MFCD00150754