Ammonium sulphate, Ultrapure
About this item
May be used for the precipitation or fractionation of proteins or for purification of antibodies. Useful for crystallographic analysis of nucleic acids and proteins.
- CAS Number:7783-20-2
- Formula:(NH₄)₂SO₄
- MDL Number:MFCD00003391
- Molecular Weight:132.14 g/mol
- Merck Index:13,00559
- Melting Pt:280°C
- Melting Pt:280 °C
- Density:1.78 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-984-1
- REACH:01-2119455044-46
3 Options Available Below
Product Details & Documents
May be used for the precipitation or fractionation of proteins or for purification of antibodies. Useful for crystallographic analysis of nucleic acids and proteins.
- CAS Number:7783-20-2
- Formula:(NH₄)₂SO₄
- MDL Number:MFCD00003391
- Molecular Weight:132.14 g/mol
- Merck Index:13,00559
- Melting Pt:280°C
- Melting Pt:280 °C
- Density:1.78 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-984-1
- REACH:01-2119455044-46
Documents
Certificates
Search by Lot NumberVWR Life Science Biochemical and Molecular Biology Reagents
Documentación del producto
Specifications for Product Family
Specification Test Results
- Arsenic:0.0001%
- Chloride:= 0.0005%
- DNase:none detected
- Heavy Metals (as Pb):= 0.0002%
- Insolubles:= 0.005%
- Iron:= 0.0005%
- Magnesium Test:Pass
- pH (5%, Water) @25°C:5 - 6
- Phosphate:= 0.0005%
- Protease:none detected
- Purity (Dry Basis):= 99.5%
- Residue on Ignition:= 0.005%
- RNase:none detected
- Selenium:= 0.002%
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- HDPE bottle1 kgEach (1kg)$977.10Case de 4 (1kg)$1,065.04Paleta de 240 (1kg)$234,503.47Supplier Number: 0191-1KG
- Supplier Number: 0191-5KG
- Supplier Number: 0191-10KG
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



