Order Entry
Mexico
ContactUsLinkComponent
SP Bel-Art Poxygrid® 250 ml Centrifuge Tube Rack, Bel-Art Products, a part of SP
SP Bel-Art Poxygrid® 250 ml Centrifuge Tube Rack, Bel-Art Products, a part of SP
  60916-260
undefined
SP Bel-Art Poxygrid® 250 ml Centrifuge Tube Rack, Bel-Art Products, a part of SP
  60916-260
 :  F19856-0250
  • Description:
    Poxygrid® Samplit™ rack holds 6 containers
  • Environmentally Preferable:
  • Cat. no.:
    60916-260
  • L×W×H:
    20.8×13.5×10.2 cm (8.2×5.3×4")
  • Supplier No.:
    198560250
Rack holds large conical and round-bottom centrifuge tubes that cannot stand by themselves (such as Falcon®, Corning®, Nunc®, and Kimble®).
  • Epoxy-coated steel wire construction
  • Coating is chemically resistant
  • Great temperature resistance
  • Autoclavable
The epoxy-coated steel wire rack is 10.2 cm (4") high and has two levels of square grids to hold 6 tubes up to 250 mL (60 mm diameter).
Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids, and weak alkalis. It will withstand a temperature range of –223 to 121°C (–369 to 250°F), and can be autoclaved.