Prefilter for Masks, 3M™
Supplier: GRAINGER,S.A. DE C.V. MX
Jump to:
About this item
For use with 5000-series respirators.
- NIOSH 42 CFR 84 approved
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
3 Options Available Below
Product Details & Documents
For use with 5000-series respirators.
- NIOSH 42 CFR 84 approved
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Product Family Options
Product Information
- Approved For Use AgainstAvailabilityPrice
- 56222-931P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators.P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators.Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respiratorsPaquete de 2$15,360.00Case de 50 (2UN)$13,004.70Supplier Number: 2091Specifications:MarketsourceMarketsource
- 56621-940P100 filter "organic vapor odors, solid and liquid particles including oil" for use with 3M™ 5000, 6000, 7000 series respiratorsP100 filter "organic vapor odors, solid and liquid particles including oil" for use with 3M™ 5000, 6000, 7000 series respiratorsRemoves organic vapor odors below the OSHA PELs, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, 7000 series respiratorsPaquete de 2$370.00Case de 50 (2UN)$499.10Supplier Number: 7000029657
- 56221-054N95 filter for use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826N95 filter for use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826For use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826Paquete de 10$586.00Case de 10 (10UN)$791.46Supplier Number: 7000002008
- 56222-826Prefilter retainer for use with 56221-054 prefilter.Prefilter retainer for use with 56221-054 prefilter.Discontinued-Supplier Number: -