Order Entry
Mexico
ContactUsLinkComponent
Acrodisc® Sterile Syringe Filters, Pall® Life Sciences
 :  Cytiva
undefined
Acrodisc® Sterile Syringe Filters, Pall® Life Sciences
 :  Cytiva
  • Catalog No:
  • 28143-298
  • 28143-307
  • Type:
  • Fluorodyne® II
  • Posidyne®
  • Max. operating pressure:
  • 5.4 bar at ambient temperature
  • 5.4 bar at ambient temperature
  • Max. temperature:
  • 60 °C at 4.1 bar
  • 60 °C at 4.1 bar
  • Water flow rate:
  • 130 ml/min. at 2.1 bar (30 psi)
  • 77 ml/min. at 2.1 bar (30 psi)
Frequently Bought Together