Order Entry
Mexico
ContactUsLinkComponent
 
PYREX® Resin Reaction Kettles with Covers, Corning
  36390-042
 :  Corning
Product available on GSA Advantage®
undefined
PYREX® Resin Reaction Kettles with Covers, Corning
  36390-042
 :  Corning
 :  6947-1L
Product available on GSA Advantage®

 

  • Capacity:
    1 L
  • Description:
    Reaction kettle
  • Cat. no.:
    36390-042
  • O.D.×H:
    108 x 220 mm with lid
  • Supplier No.:
    6947-1L

 

 

PYREX® borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

These vessels have lids with openings to accommodate four Standard Taper joints. Interchangeable Standard Taper joints allow quick assembly with condensers, stirrers and funnels.

Flanges of lids and vessels are finely ground for a tight seal.

For use with heating mantle only.