Order Entry
Mexico
ContactUsLinkComponent
8233 Disposable N100 Respirators, 3M™
8233 Disposable N100 Respirators, 3M™
Catalog # 56621-928
undefined
8233 Disposable N100 Respirators, 3M™
Catalog # 56621-928
Supplier Number:  7000002028
Specifications
  • Description:
    8233 N100 Respirator
  • Environmentally Preferable:
  • Cat. no.:
    56621-928
  • Supplier No.:
    8233
Specifications
About this item
This respirator is well suited for those who want NIOSH's highest rated filtration efficiency in a maintenance-free respirator.
  • NIOSH’s highest rated filtration efficiency in maintenance-free respirator
  • For grinding, welding, soldering, torch cutting, metal pouring, and OSHA substance-specific particle exposures including lead, cadmium, and arsenic
  • Adjustable straps, Cool Flow® exhalation valve, and conformable face-seal makes this a comfortable, durable respirator
  • Provides minimum filtration efficiency of 99.97% against solid and liquid particles that do not contain oil
Can be used for grinding, welding, soldering, torch cutting, metal pouring, and OSHA substance-specific particle exposures including lead, cadmium, and arsenic. Adjustable straps, Cool Flow® exhalation valve, and a conformable face seal make this a comfortable, durable respirator. Provides minimum filtration efficiency of 99.97% against solid and liquid particles that do not contain oil.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.