Order Entry
Mexico
ContactUsLinkComponent
Vacushield™ Vent Device, Cytiva (Formerly Pall Lab)
Vacushield™ Vent Device, Cytiva (Formerly Pall Lab)
Catalog # 55095-006
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Vacushield™ Vent Device, Cytiva (Formerly Pall Lab)
Catalog # 55095-006
Supplier:  Cytiva
Supplier Number:  4402
Product available on GSA Advantage®

Specifications

  • Dimensions:
    7.3 dia. X 8.2 L cm
  • Cat. no.:
    55095-006
  • Supplier No.:
    4402

Specifications

About this item

This disposable protective device is designed to prevent aqueous liquid and aerosol contamination of vacuum pumps or aspiration suction systems.

  • Disposable
  • High air flow
  • High bacterial retention

It provides high air flow rates and bacterial retention without loss of pump efficiency. Non-sterile. May be autoclaved or ETO sterilized. Hydrophobic, microporous PTFE membrane filter achieves 99.97% D.O.P. retention (0.3 µm particles in air). Features polypropylene housing and 6.4 - 12.7 mm (¹/₄ - ¹/₂") stepped hose barb connections (internal taper accepts male Luer fitting). Dimensions: 7.3 Ø × 8.2 L cm (2⁷/₈×3¹/₄").