KIMAX® Capillary Melting Point Tubes, Kimble Chase,DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Capillary tubes available in 100 mm length with both ends open, or 90 mm length with one end sealed.
Tubes 36982-008 and 36984-003 have an O.D. of approx. 1.5 to 1.8 mm with a 0.2 mm wall thickness.
Tubes 36983-000 and 36984-025 comply with requirements given in the United States Pharmacopoeia (USP) and have an I.D. of approx. 0.8 to 1.1 mm with a 0.25 mm wall thickness.
Delivery: Packed in vials of 100 tubes, 20 vials per case.