Gas Diffuser Stone, Saint-Gobain
Catalog # 32573-007
Supplier: SAINT GOBAIN ABRASIVES
CAS Number:
Specifications
- Description:Gas Diffuser Stone
- Environmentally Preferable:
- Cat. no.:32573-007
- Supplier No.:3590001A
Specifications
About this item
Spherical 2.5 cm (1") fine porous aerator stone made of Alundum® fused crystalline aluminum oxide.
Delivery: Supplied with a cadmium-plated brass nipple fitting serrated for 0.5 cm (3/16") rubber tubing.