About this item
These ultra-sharp blades section specimens without striations, distortion or chattering. Low-profile blades fit all Accu-Edge® blade holders for microtomy and cryotomy. High-profile blades are compatible with holders designed for disposable blades 14mm in height. Blades come in dispensers that hold 50 blades and deliver them in seconds, automatically ejecting the used blade.
Low-profile blades fit all Accu-Edge® blade holders for microtomy and cryotomy. High-profile blades are compatible with holders designed for disposable blades 14mm in height. Blades come in dispensers that hold 50 blades and deliver them in seconds, automatically ejecting the used blade.