Order Entry
Mexico
ContactUsLinkComponent
 
Supelco® Reflectoquant® Formaldehyde Test, MilliporeSigma
  EM1.16989.0001
 :  
Supelco® Reflectoquant® Formaldehyde Test, MilliporeSigma
  EM1.16989.0001
 :  1.16989.0001
 :  

 

  • Chemical method:
    Reflectometric
  • Pack type:
    Plastic box
  • Description:
    Formaldehyde Test
  • Size:
    1 pc
  • Storage temperature:
    15…25 °C
  • Cat. no.:
    EM1.16989.0001
  • Measurement Range:
    1.0 - 45.0 mg/L
  • Supplier No.:
    1.16989.0001
  • No. of Tests:
    50

 

 

The Reflectoquant Formaldehyde test includes test strips and all necessary additional reagents. Formaldehyde is used as a disinfectant for surfaces, medical equipment and also as a raw material in binding agents, fixing agents or as a preservative. Formaldehyde reacts with a Triazole to form a purple-red tetrazine that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the formaldehyde concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for formaldehyde. Further quantitative determination methods can be found with our Spectroquant tests.

For use with REFLECTOQUANT® RQflex® 10 Plus, RQflex® 10, RQflex® 20.