Nunc® Centrifuge Tubes with EZ Flip™ Cap, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Conical-bottom, graduated tubes feature an ergonomically designed hinged cap that allows one-handed opening and closing.
- Flat, leakproof caps prevent cross contamination
- Tubes feature a white writing area for sample identification
Clear polypropylene tubes are chemical resistant and have high speed tolerances, allowing them to be used for a wide range of applications.
Related Information
Certificates
Search by Lot NumberThermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Frequently Bought Together