Microhematocrit Tube Reader, McCormick Scientific
Catalog # 15176-816
Proveedor: Avantor
Especificaciones
- Description:Microhematocrit Tube Reader
- Cat. no.:15176-816
- Supplier No.:39111004
Especificaciones
Acerca de este artículo
Reader is constructed of durable, washable, vinyl-laminated plastic and features two ruled charts for precise determination of values.
- Durable
- Washable
- Vinyl-laminated plastic construction
Two ruled charts are provided to allow precise determination of packed cell volumes.