Order Entry
Mexico
ContactUsLinkComponent
KIMAX® Glass Pennyhead [ST] Stoppers, Flask Length, Kimble Chase
KIMAX® Glass Pennyhead [ST] Stoppers, Flask Length, Kimble Chase
Catalog # 89002-174
undefined
KIMAX® Glass Pennyhead [ST] Stoppers, Flask Length, Kimble Chase
Catalog # 89002-174
Supplier Number:  41900R-13
Specifications
  • Description:
    Solid stopper
  • Environmentally Preferable:
  • Cat. no.:
    89002-174
  • ST Stopper No.:
    13
  • Supplier No.:
    41900R-13
Specifications
About this item
Stoppers are for use with flasks, mixing cylinders, separatory funnels, and other glassware
Designed from ASTM Specification E675.
Size 13 stopper is solid. Size 38 stopper is hollow.