Super Erecta Shelf® Lab Security Cart, Metro™
Catalog # 82000-940
Supplier: Metro
Specifications
- Width:24"
- Height:39"
- Length:30"
- Cat. no.:82000-940
- Shipping Weight:85 lbs.
- Load Capacity:600 lbs. evenly distributed
- Supplier No.:SECMLAB
Specifications
About this item
Stainless steel security cart with flip-up door provides secure storage for animal cage, lab instrument, or hazardous chemical transport
Features two stainless steel wire shelves, one stainless steel security module, and one stainless steel solid shelf that can be used as a work surface and features a raised ship's edge to contain unwanted spills. Super Erecta Shelf® construction offers ease of assembly, absolute rigidity, and quick and simple relocation of wire shelves. Open construction minimizes dust accumulation and maximizes visibility and air circulation.
Ordering information: Cart includes four donut bumpers and four 12.7cm (5") polyurethane casters (2 swivel, 2 brake).