Order Entry
Mexico
ContactUsLinkComponent
Tissue-Tek® Embedding Ring Filing Cabinet, Sakura® Finetek
Tissue-Tek® Embedding Ring Filing Cabinet, Sakura® Finetek
Catalog # 25608-836
undefined
Tissue-Tek® Embedding Ring Filing Cabinet, Sakura® Finetek
Catalog # 25608-836
Supplier Number:  4169
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Cabinet with 1000 white embedding rings
  • Cat. no.:
    25608-836
  • Supplier No.:
    4169
Specifications
About this item
Provides permanent storage/reference for blocks embedded in Tissue-Tek® embedding rings. Stackable cabinets accommodate 1000 rings. One-piece construction eliminates hinges and doors for dust-free storage. Includes six, impact-resistant drawers, each with an identification area. Dimensions: 24.1Wx43.2Dx38.1Hcm (91/2x17x15").
  • Includes six, impact-resistant drawers, each with an identification area
  • Dimensions: 24.1 W x 43.2 D x 38.1 H cm (91/2 x 17 x 15")
Stackable cabinets accommodate 1000 rings. One-piece construction eliminates hinges and doors for dust-free storage.