Peel-A-Way® Disposable Embedding Molds
Product Details & Documents
Disposable paraffin molds are designed to "Peel-A-Way" from the solidified embedded block. For plastic or paraffin embedding. Eliminates need for release agent, additional parts, and block trimming. Inside fingers hold ID slip, which becomes embedded in block for positive, permanent identification.
For plastic or paraffin embedding. Eliminates need for release agent, additional parts, and block trimming. Inside fingers hold ID slip, which becomes embedded in block for positive, permanent identification.
Specifications
- Description:Truncated Mold, Truncated from 22×22 mm
- L×W×D:12×12×20 mm (15/32 × 15/32 × 25/32")