LITE-DRI® Absorbent, New Pig
Catálogo # 46000-496
Proveedor: 2 D 2 S.A. DE C.V. MX
Especificaciones
- Description:Absorbs Oils, Coolants, Solvents, and Water
- Size:22 lb.
- Cat. no.:46000-496
- Packaging:BAGA
- Supplier No.:PLP201
Especificaciones
Acerca de este artículo
Unlike clay, which is simply coated with liquid, fast-wicking recycled cellulose actually soaks up the spill the moment it touches
- Absorbs and retains oils, coolants, solvents and water
- Bag features a built-in handle for easy portability, handling and dispersal
- Recycled cellulose is ideal when green products are desired or required
- Can be incinerated after use to reduce waste or for fuels blending
Three times more absorbent means you'll use less material on the spill, reducing your labor and disposal costs. Silica-free composition is non-abrasive and does not pose the health hazards associated with breathing in clay dust. Non-abrasive composition won't harm finished floors or damage expensive machinery. Light-weight absorbent is easier than clay to carry and use without injury.