Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Protective Labels

VWR® Protective Labels

Write-on hazard warning labels, 24×50.8 mm (0.94×2")
Catalog # 89097-892
Supplier:   VWR International
VWR® Protective Labels
Catalog # 89097-892
Supplier:  VWR International
VWR® Protective Labels
VWR® Protective Labels

Write-on hazard warning labels, 24×50.8 mm (0.94×2")

Price
$132.20
Log in for availability
Case (1UN)
Product Details & Documents

Radiant-orange, write-on labels with a protective cover prevent the loss of valuable information. Designed for labeling reagents, samples, or noting calibration data. Simply write information on label, peel backing from transparent film, and press film over the label. Apply the label to any clean, dry surface. No dispenser required.

  • No dispenser required
  • Easy and simple application

Designed for labeling reagents, samples, or noting calibration data.

Labels protect written information from tampering, moisture, smearing, heat and cold, chemicals, and solvents.

Simply write information on label, peel backing from transparent film, and press film over the label. Apply the label to any clean, dry surface.

Packaging: 500 labels per roll.

Specifications
  • Description:
    Write-on hazard warning labels
  • Dimensions:
    24×50.8 mm (0.94×2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.