Disposable Inoculating Loops and Needles, Sterile, BD Difco™
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Certificates
About this item
Made of specially formulated plastic resin
Flexible enough to prevent gouging agar yet firm enough for picking difficult colonies. Comfortable, smooth-textured, flat handle. Color-coded for easy size identification.
Ordering information: Packaged sterile, ready to use, in tamper-evident, resealable clear plastic tubes.