Order Entry
Mexico
ContactUsLinkComponent
Synthware Kuderna-Danish Flasks, Kemtech America
Synthware Kuderna-Danish Flasks, Kemtech America
Catalog # 60002-256
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Kuderna-Danish Flasks, Kemtech America
Catalog # 60002-256
Supplier Number:  F984500
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Capacity:
    500 ml
  • Cat. no.:
    60002-256
  • Supplier No.:
    F984500
Specifications
About this item
Top outer [ST] joint size: 24/40.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Kuderna-Danish style
Lower inner [ST] joint size: 19/22.