Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Long-Stem Thermometers
VWR® Long-Stem Thermometers
VWR® Long-Stem Thermometers
VWR® Long-Stem Thermometers
Measure sample temperature in hard-to-reach areas, including cuvettes, test tubes, flasks, and beakers.
  • Traceable®
  • Two temperature ranges available
  • Two lengths available
  • Stainless steel probe
  • Read °C and °F
Also read temperature in water baths, foods, gases, and semisolids. Simply flip a switch to change the 6.4mm (1/4") high LCD display between °C and °F. Readings are updated every second. Safer than mercury thermometers due to plastic and stainless steel construction. Stainless steel probe is resistant to acids, bases, solvents, and most laboratory chemicals.
 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Supplied with a 1.5V silver-oxide battery and a protective case that can also be used as a holder.
 : VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
  • Catalog No:
  • 61220-416
  • 23609-176
  • 23226-658
  • 23609-174
  • Temperature resolution:
  • 0.1° from –20 to 200°C
  • 0.1° from –20 to 200°C
  • 0.1° from –20 to 200°C
  • 0.1° from –20 to 200°C
  • Probe:
  • 20.3 cm (8")
  • 28.9 cm (11⅜")
  • 20.3 cm (8")
  • 28.9 cm (11⅜")
Product Family Options
Thermometers
  • Description
    Range
    Accuracy
    Probe
    Length
    Ø
  • Standard Thermometer
    –50...150 °C (–58...302 °F)
    ±1 °C between –30...+150 °C
    20.3 cm (8")
    27.9 cm (11")
    20.6 mm (13/16")
    Each (1UN)
    Each (1UN) - $1,217.07
    $1,217.07
    Each (1UN)
       $1,217.07
    Specifications
    Cat. No.
    61220-416
    Temperature resolution
    0.1° from –20 to 200°C
    Probe
    20.3 cm (8")
  • Standard Thermometer
    –50...300 °C (–58...572 °F)
    ±1 °C between –30...+150 °C
    28.9 cm (11⅜")
    36.5 cm (14⅜")
    20.6 mm (13/16")
    Each (1UN)
    Each (1UN) - $1,530.82
    $1,530.82
    Each (1UN)
       $1,530.82
    Specifications
    Cat. No.
    23609-176
    Temperature resolution
    0.1° from –20 to 200°C
    Probe
    28.9 cm (11⅜")
  • ULTRA™ Thermometer
    –50...150 °C (–58...302 °F)
    ±0.2 °C at tested points 0° and 50 °C only (all other points ±1 °C)
    20.3 cm (8")
    27.9 cm (11")
    20.6 mm (13/16")
    Each (1UN)
    Each (1UN) - $3,161.39
    $3,161.39
    Each (1UN)
       $3,161.39
    Specifications
    Cat. No.
    23226-658
    Temperature resolution
    0.1° from –20 to 200°C
    Probe
    20.3 cm (8")
  • ULTRA™ Thermometer
    –50...300 °C (–58...572 °F)
    ±0.5 °C (at tested points 0 and 100 °C; all other points ±1 °C)
    28.9 cm (11⅜")
    36.5 cm (14⅜")
    20.6 mm (13/16")
    Each (1UN)
    Each (1UN) - $2,318.30
    $2,318.30
    Each (1UN)
       $2,318.30
    Specifications
    Cat. No.
    23609-174
    Temperature resolution
    0.1° from –20 to 200°C
    Probe
    28.9 cm (11⅜")