Kahnetics® Tapered Needles, Cooper Tools
Supplier: APEX TOOL GROUP
Certificates
About this item
All-polyethylene dispensing tips
Tapered design reduces dripping. Polyethylene material won’t scratch delicate surfaces. Overall length: 38.1mm (11/2").
Ordering information: Tips are packaged 50 per pack.
Frequently Bought Together