Order Entry
Mexico
ContactUsLinkComponent
Polypropylene Screw Caps, Linerless, Kimble Chase
Polypropylene Screw Caps, Linerless, Kimble Chase
  66010-682
undefined
Polypropylene Screw Caps, Linerless, Kimble Chase
  66010-682
 :  73805B-18415
  • Cap Size:
    18-415
  • Color:
    Natural
  • Cat. no.:
    66010-682
  • Supplier No.:
    73805B-18415
Polypropylene caps feature one-piece construction with a seal achieved by a unique interior design.
Caps withstand repeated autoclaving when loosened or removed from tubes.
 : Designed from ASTM Specification E982