Order Entry
Mexico
ContactUsLinkComponent
Dura Shoe Covers
  21923-220
undefined
Dura Shoe Covers
  21923-220
 :  10135-109-02
  • Shoe size:
    Universal
  • Garment application:
    Laboratory
  • Color:
    Lilac
  • Cat. no.:
    21923-220
  • Supplier No.:
    10135-109-02
Covers made of spunbound polypropylene bound to an exterior layer of compressed polyethylene (CPE) film
Seamless design effectively contains particulates from street shoes. Heavily textured and snug fitting for better slip resistance. Dimensions: 40.6Lx16.5Hcm (16x7"). Relaxed opening: 12.7cm (5"). Stretched opening: 27cm (105/8").