Order Entry
Mexico
Orders LinkContactUsLinkComponent
Microcentrifuge Tubes
Microcentrifuge Tubes
Microcentrifuge Tubes
Supplier:  BIO PLAS INC.
Certificates
Microcentrifuge Tubes
Microcentrifuge Tubes
2 Options Available Below
Product Details & Documents
Ideal for applications in freezer storage, autoclaving, boiling, or irradiation.
  • External ridges fit rack serrations and allow one-handed opening
Colored PPCO screw caps are fitted with an ethylene propylene O-ring for reliable, leakproof performance. Compatible with most microcentrifuges with fixed angle heads. Withstands over 20000×g.
Tubes are made of reusable polypropylene copolymer (PPCO). Seamless conical construction with squared external threads prevents hang up of samples and reduces contamination.
Ordering information: Dual-ended polypropylene tool 20170-688 provides a combination of socket wrench and bottle top action for quick, effective tube access. Tubes and caps are available separately.
Product Family Options
Tubes
Product Information
  • Volume
    Color
    Maximum RCF
    Style
    Graduations
    Availability
    Price
  • 20170-723
    Screw cap microcentrifuge tube
    Screw cap microcentrifuge tube
    0.5 ml
    Amber
    20000×g
    Skirted
    No
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $1,744.69
    $1,744.69
    Paquete de 500
       $1,744.69
    Supplier Number: 4201
Screw Caps
Product Information
  • Volume
    Color
    Maximum RCF
    Style
    Graduations
    Availability
    Price
  • 20170-793
    Screw cap with O-ring
    Screw cap with O-ring
    Amber
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $2,538.41
    $2,538.41
    Paquete de 500
       $2,538.41
    Supplier Number: 4224R