Order Entry
Mexico
ContactUsLinkComponent
Sani-Cloth® Plus Germicidal Disposable Wipes, PDI®
  15648-990
undefined
Sani-Cloth® Plus Germicidal Disposable Wipes, PDI®
  15648-990
 :  PDI Q89072
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Cat. no.:
    15648-990
  • Packaging:
    160 Wipes/Canister
  • W×L:
    15.2 x 17 cm (6 x 63/4")
  • Supplier No.:
    Q89072
Deep well lid provides ample space to store and access next wipe. Prominent contact time for easy identification and improved compliance with accrediation standards. Categorization of pathogens allows for quick identification of relevant efficacy claims. Safety icon clearly communicates key safety information on both label and lid.
  • EPA-registered, low-level disinfectant
  • Pre-moistened and ready-to-use
  • Cleans and disinfects in one step
  • Bactericidal and virucidal
  • Compatible with a broad range of hard nonporous surfaces
  • Reduces waste and saves time
 : Meets CDC, OSHA, AHE and CMS Tag F441 guidelines.