Order Entry
Mexico
ContactUsLinkComponent
Scienceware® Biohazard Disposal Can with Lift-up Cover, Bel Art
Scienceware® Biohazard Disposal Can with Lift-up Cover, Bel Art
Catalog # 74950-062
undefined
Scienceware® Biohazard Disposal Can with Lift-up Cover, Bel Art
Catalog # 74950-062
Supplier Number:  F13197-0000

Specifications

  • Dimensions:
    22.9D x 30.5W x 29.2H cm (9 x 12 x 111/2")
  • Cat. no.:
    74950-062
  • Supplier No.:
    131970000

Specifications

About this item

Bright red safety disposal container is suitable for biological/biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement. Molded of polypropylene for long service life, this can is puncture-resistant and autoclavable at 121°C (250°F). One piece hinged lid protects users from contact with the contents, helps retain odors and contains airborne contaminants.

  • Polypropylene construction
  • Long service life
  • Puncture-resistant
  • Autoclavable
  • One-piece hinged lid contains odors and protects users from contents

Molded of polypropylene for long service life, this can is puncture-resistant and autoclavable at 121°C (250°F). One-piece hinged lid protects users from contact with the contents, helps retain odors and contains airborne contaminants.

19 x 23" autoclave bags (47750-748 or 47750-758) fit perfectly and are secured with an internal stainless steel wire bar.

Ordering information: Bags sold separately.