- Capacity:37.9 L (10 gal.)
- Dimensions:39.3W x 61D x 65.4H cm (151/2 x 24 x 253/4")
- Environmentally Preferable:
- Cat. no.:95041-442
- Supplier No.:31143665
These large red containers are ideal for the convenient disposal of large instruments and high volumes of waste. The clear lid keeps the contents and fill level visible for increased safety. Containers feature molded carrying handles to aid in lifting heavy containers for final disposal. Container 82018-056 has a split lid with an integrated sharps port.
Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
The clear lid keeps the contents and fill level visible for increased safety. Containers feature molded carrying handles to aid in lifting heavy containers for final disposal. Container 82018-056 has a split lid with an integrated sharps port.