Order Entry
Mexico
ContactUsLinkComponent
Sharps Disposal Containers, Large Volume, Covidien
  58578-090
undefined
Sharps Disposal Containers, Large Volume, Covidien
  58578-090
 :  8938
  • Capacity:
    68.1 L (18 gal.)
  • Description:
    With Clear Lid
  • Dimensions:
    66H x 46.4W x 32.4D cm (26 x 181/4 x 123/4")
  • Cat. no.:
    58578-090
  • Supplier No.:
    8938
Red containers offer both large and small openings for infectious and biohazardous waste of various types. Containers with hinged lids can easily accommodate the largest sharps suction canisters, trocars, endoscopic instruments, and more. Hinged lids permit temporary closure when not in use. All containers lock for final disposal. Containers 82003-550 and 15704-216 feature a hinged, sealing gasket lid to provide leak-resistant containment of contents. Container 89004-914 features a gasketed, slide-top lid. It is PG II rated and DOT approved for transport of regulated medical waste including cultures and stocks.
Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
Containers with hinged lids can easily accommodate the largest sharps suction canisters, trocars, endoscopic instruments, and more. Hinged lids permit temporary closure when not in use. All containers lock for final disposal. Containers 82003-550 and 15704-216 feature a hinged, sealing gasket lid to provide leak-resistant containment of contents. Container 89004-914 features a gasketed, slide-top lid. It is PG II rated and DOT approved for transport of regulated medical waste including cultures and stocks.