Flux and Liquid Dispensers, Plato™, ITW Chemtronics
15551-500
- Capacity:60 ml (2 oz.)
- Description:Cut-to-Size Dispenser, Capillary I.D. 0.13–0.9 mm
- Cat. no.:15551-500
- Supplier No.:FD-21
POS–A–LOC® leakproof liquid dispensers with needles conveniently dispense specific amounts of a variety of liquids without dripping, spilling, or waste.
Stainless steel replacement needles also available.
The cut-to-size flux dispenser allows you to cut the tip opening to the correct size.