Order Entry
Mexico
ContactUsLinkComponent
 
Cyclohexane, anhydrous ≥99.8%, DriSolv®, Supelco®
Catalog # EM-CX2284-6
CAS Number:  
undefined
Cyclohexane, anhydrous ≥99.8%, DriSolv®, Supelco®
Catalog # EM-CX2284-6
Supplier Number:  CX2284-6
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, septum-sealed
  • Size:
    1 L
  • Environmentally Preferable:
  • Cat. no.:
    EM-CX2284-6
  • Supplier No.:
    CX2284-6

Specifications

Specification Test Results

  • Assay (GC):
    99.5% min
  • Appearance:
    Clear liquid, free from particulates
  • Color (APHA):
    10 max
  • Filtered through 0.2 µm filter:
    To pass test
  • Residue after evaporation:
    5ppm max
  • Water (H2O):
    50ppm max

Specification Test Results

About this item

Synonyms: Hexahydrobenzene, Hexamethylene
 :
 :84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Flash Pt:-18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
 :Packed in septum-sealed bottle.
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814