PYREX® Short Neck Boiling Flask, Round Bottom, [ST] Joint, Corning
Supplier: Corning
Certificates
About this item
Boiling flasks with full length [ST] outer joints are manufactured from borosilicate glass.
Normal working temperature is 230 °C (446 °F). Heavy wall flasks feature walls that are 25-30% thicker than standard walls, for increased mechanical strength.
Certifications: Flasks 89090-694 and 89090-884 reference ASTM E123.
Specifications
- Catalog No:
- 89090-798
- 89090-884
- 32645-028
- 89090-862
- 89091-464
- 89090-694
- 22108-588
- 89090-618
- 89090-616
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey