Tetramethylammonium hydroxide in methanol 24.5-25.5%
Catalog # JT5879-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle, pressure bottom
- Size:8.5 lbs.
- Cat. no.:JT5879-3
- Supplier No.:5879-03
Specifications
Specification Test Results
- Assay (C₄H₁₃NO) (by acid-base titration):24.5 - 25.5 %
- Carbonate (CO₃):≤100 ppm
- Color (APHA):≤10
- Halide (as Cl):≤5 ppm
- Trace Impurities - Aluminum (Al):≤3.5 ppb
- Trace Impurities - Barium (Ba):≤2.0 ppb
- Trace Impurities - Boron (B):≤10.2 ppb
- Trace Impurities - Boron (FUT):≤10.2 ppb
- Trace Impurities - Boron (B) (Sub-supplier):≤2.0 ppb
- Trace Impurities - Calcium (Ca):≤7.5 ppb
- Trace Impurities - Chromium (Cr):≤2.0 ppb
- Trace Impurities - Cobalt (Co):≤1.3 ppb
- Trace Impurities - Copper (Cu):≤2.0 ppb
- Trace Impurities - Iron (Fe):≤2.7 ppb
- Trace Impurities - Lead (Pb):≤1.4 ppb
- Trace Impurities - Lithium (Li):≤2.0 ppb
- Trace Impurities - Magnesium (Mg):≤1.7 ppb
- Trace Impurities - Manganese (Mn):≤1.7 ppb
- Trace Impurities - Nickel (Ni):≤1.3 ppb
- Trace Impurities - Potassium (K):≤2.8 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤5.6 ppb
- Trace Impurities - Tin (Sn):≤1.7 ppb
- Trace Impurities - Titanium (Ti):≤1.9 ppb
- Trace Impurities - Zinc (Zn):≤2.3 ppb
- Particle Count at point of fill - 1.0 µm and greater:≤25 par/ml
- Particle Count at point of fill - 0.5 µm and greater:≤400 par/ml
Specification Test Results
About this item
:
:91.15 g/mol
ADR:8,II
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:200-882-9
UN:1835
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280