Order Entry
Mexico
ContactUsLinkComponent
 
Silver nitrate (0.0058 - 0.058 mol/l; 0.0058 - 0.058 N) in aqueous solution 0.0141 M analytical reagent
Catalog # RC686032
Supplier:  RICCA CHEMICAL CO
Small Business Enterprise
CAS Number:  
undefined
Silver nitrate (0.0058 - 0.058 mol/l; 0.0058 - 0.058 N) in aqueous solution 0.0141 M analytical reagent
Catalog # RC686032
Supplier:  RICCA CHEMICAL CO
Supplier Number:  R6860000-1B
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, amber
  • Size:
    1 L
  • Cat. no.:
    RC686032
  • Supplier No.:
    R6860000-1B

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay at 20 °C (traceable to NIST SRM 999):
    0.01410 ± 0.00002 N at 20.0 °C

Specification Test Results

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414