
Sodium hydroxide (5 - 60%) in aqueous solution 2.5 N BAKER ANALYZED®
Catalog # JT5666-2
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-Regulated, With InformationCertificates
Specifications
- Pack type:PE bottle, narrow mouth, 32 oz.
- Size:1 L
- Cat. no.:JT5666-2
- Supplier No.:5666-02
Specifications
Specification Test Results
- Normality:2.40 - 2.60
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Boiling Pt:119 - 135 °C (1013 hPa)
Melting Pt:9…10 °C
Melting Pt:9 - 10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:Management of Change (MOC) category = NI
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548