Sodium hydroxide ( 0.13 mol/l; 0.13 N) in aqueous solution 0.01 N BAKER ANALYZED®
Catalog # JT5663-2
Supplier: AVANTOR PERFORMANCE MATERIALS MX
CAS Number:
MOC: Non-Regulated, With InformationCertificates
Specifications
- Pack type:PE bottle, narrow mouth
- Size:1 L
- Cat. no.:JT5663-2
- Supplier No.:5663-02
Specifications
Specification Test Results
- Normality:0.0095 - 0.0105
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
Specification Test Results
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,III
:Management of Change (MOC) category = NI
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548